Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419331.6 | XP_419331.1 | 1 - 2 | 140 | 123 | |
GenBank mRNA | BX931436.2 | - | 1 - 2 | 121 | 123 | |
BX932844.2 | - | 1 - 2 | 122 | 123 | ||
BX933982.1 | - | 1 - 2 | 131 | 123 | ||
BX934618.1 | - | 1 - 2 | 134 | 123 | ||
BX935753.1 | - | 1 - 2 | 128 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |