Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025149345.1 | XP_025005113.1 | 5 - 6 | 839 | 139 | |
XM_419728.6 | XP_419728.3 | 6 - 7 | 970 | 139 | ||
GenBank mRNA | BX935610.1 | - | 5 - 6 | 1003 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |