Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_414755.5 | XP_414755.2 | 8 - 9 | 1749 | 113 | |
GenBank mRNA | BX934565.2 | - | 6 - 7 | 911 | 113 | |
BX936040.1 | - | 6 - 7 | 919 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |