Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015296644.1 | XP_015152130.1 | 2 - 3 | 494 | 69 | |
XM_015296645.2 | XP_015152131.1 | 2 - 3 | 354 | 69 | ||
XM_015296646.2 | XP_015152132.1 | 2 - 3 | 357 | 69 | ||
XM_015296647.2 | XP_015152133.1 | 2 - 3 | 495 | 69 | ||
XM_025142311.1 | XP_024998079.1 | 2 - 3 | 480 | 69 | ||
XM_025142312.1 | XP_024998080.1 | 2 - 3 | 495 | 69 | ||
GenBank mRNA | BX931341.2 | - | 1 - 2 | 96 | 69 | |
BX936120.1 | - | 2 - 3 | 337 | 69 | ||
CR405816.1 | - | 2 - 3 | 287 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |