Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416439.6 | XP_416439.3 | 10 - 11 | 1443 | 62 | |
GenBank mRNA | BX950662.1 | - | 10 - 11 | 1121 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |