Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416439.6 | XP_416439.3 | 13 - 14 | 1948 | 97 | |
GenBank mRNA | BX950662.1 | - | 13 - 14 | 1626 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |