Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416439.6 | XP_416439.3 | 2 - 3 | 600 | 96 | |
GenBank mRNA | BX950662.1 | - | 2 - 3 | 278 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |