Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416439.6 | XP_416439.3 | 3 - 4 | 679 | 108 | |
GenBank mRNA | BX950662.1 | - | 3 - 4 | 357 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |