Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302168.1 | NP_001289097.1 | 3 - 4 | 823 | 77 | |
GenBank mRNA | BX929857.2 | - | 3 - 4 | 833 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |