Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERRFI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015297000.2 | XP_015152486.1 | 4 | 1002 | 117 | |
XM_015297001.2 | XP_015152487.1 | 5 | 1439 | 117 | ||
XM_025142735.1 | XP_024998503.1 | 4 | 959 | 117 | ||
GenBank mRNA | BX932996.2 | - | 1 | 650 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon |