Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_415246.6 | XP_415246.1 | 3 - 4 | 588 | 106 | |
GenBank mRNA | BX934413.2 | - | 3 - 4 | 570 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |