Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_415246.6 | XP_415246.1 | 6 - 7 | 1064 | 60 | |
GenBank mRNA | BX934413.2 | - | 6 - 7 | 1046 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |