Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199512.1 | NP_001186441.1 | 2 - 3 | 135 | 121 | |
GenBank mRNA | BX259616.3 | - | 2 - 3 | 136 | 121 | |
BX936066.2 | - | 2 - 3 | 135 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |