Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015291728.2 | XP_015147214.1 | 8 - 9 | 1278 | 117 | |
XM_416426.6 | XP_416426.3 | 7 - 8 | 1084 | 117 | ||
GenBank mRNA | CR385222.1 | - | 7 - 8 | 1073 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |