Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015294419.2 | XP_015149905.1 | 4 | 1331 | 68 | |
GenBank mRNA | CR385241.1 | - | 1 | 850 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |