Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004949515.3 | XP_004949572.1 | 10 - 11 | 1153 | 79 | |
XM_004949517.3 | XP_004949574.1 | 9 - 10 | 1032 | 79 | ||
XM_015280701.1 | XP_015136187.1 | 10 - 11 | 1205 | 79 | ||
XM_015280702.1 | XP_015136188.1 | 11 - 12 | 1354 | 79 | ||
XM_015280703.2 | XP_015136189.1 | 10 - 11 | 1151 | 79 | ||
XM_015280704.2 | XP_015136190.1 | 10 - 11 | 1354 | 79 | ||
XM_025144918.1 | XP_025000686.1 | 9 - 10 | 1084 | 79 | ||
XM_424912.6 | XP_424912.3 | 10 - 11 | 1115 | 79 | ||
GenBank mRNA | CR385816.1 | - | 3 - 4 | 303 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |