Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015289700.2 | XP_015145186.1 | 5 - 6 | 1251 | 114 | |
GenBank mRNA | CR387407.1 | - | 5 - 6 | 880 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |