Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLCCI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012855.3 | NP_001012873.3 | 4 - 5 | 793 | 76 | |
XM_015281400.2 | XP_015136886.1 | 4 - 5 | 1260 | 76 | ||
GenBank mRNA | AJ721135.1 | - | 4 - 5 | 462 | 76 | |
BX930403.1 | - | 4 - 5 | 414 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |