Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPRML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025144186.1 | XP_024999954.1 | 1 | 798 | 96 | |
GenBank mRNA | BX929633.1 | - | 1 | 749 | 96 | |
BX950687.2 | - | 2 | 513 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |