Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLT8D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277383.2 | NP_001264312.2 | 9 - 10 | 1177 | 138 | |
GenBank mRNA | BU404500.1 | - | 2 - 3 | 152 | 138 | |
BX929886.1 | - | 9 - 10 | 1177 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |