Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_420773.5 | XP_420773.1 | 2 | 687 | 70 | |
GenBank mRNA | BX930084.1 | - | 1 | 344 | 70 | |
BX930139.2 | - | 1 | 340 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |