Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETV7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004934881.3 | XP_004934938.1 | 2134 | 74 | ||
XM_004934882.3 | XP_004934939.1 | 2075 | 74 | |||
XM_004934884.3 | XP_004934941.1 | 765 | 74 | |||
XM_025143802.1 | XP_024999570.1 | 936 | 74 | |||
XM_025143803.1 | XP_024999571.1 | 1968 | 74 | |||
XM_025143804.1 | XP_024999572.1 | 1970 | 74 | |||
XM_025143805.1 | XP_024999573.1 | 1911 | 74 | |||
XM_025143806.1 | XP_024999574.1 | 515 | 74 | |||
GenBank mRNA | BX950624.2 | - | 521 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus |