Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19H17orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_429523.3 | XP_429523.1 | 5 - 6 | 1090 | 84 | |
GenBank mRNA | CR390154.1 | - | 4 - 5 | 571 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |