Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Gg03324869_m1
See other RTKN2 GE Assays ›
Gene Symbol:
RTKN2
Gene Name:
rhotekin 2
Gene Aliases:
PLEKHK1, rhotekin-2
Chromosome Location:
Chr.6: 8782767 - 8954175 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID
Gg03324869_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: RTKN2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001199597.1
NP_001186526.1
9 - 10
891
67
XM_015288109.2
XP_015143595.1
8 - 9
910
67
XM_025151516.1
XP_025007284.1
8 - 9
910
67
XM_025151517.1
XP_025007285.1
8 - 9
910
67
XM_025151518.1
XP_025007286.1
8 - 9
910
67
XM_025151519.1
XP_025007287.1
8 - 9
910
67
XM_025151520.1
XP_025007288.1
8 - 9
1021
67
XM_025151521.1
XP_025007289.1
20 - 21
4128
67
XM_025151522.1
XP_025007290.1
20 - 21
12226
67
XM_025151523.1
XP_025007291.1
20 - 21
4014
67
XM_025151524.1
XP_025007292.1
21 - 22
11263
67
XM_025151525.1
XP_025007293.1
21 - 22
12360
67
XM_025151526.1
XP_025007294.1
20 - 21
12120
67
XM_025151527.1
XP_025007295.1
21 - 22
11216
67
XM_025151528.1
XP_025007296.1
19 - 20
12038
67
XM_025151529.1
XP_025007297.1
20 - 21
11156
67
XM_025151530.1
XP_025007298.1
20 - 21
2904
67
XM_025151531.1
XP_025007299.1
19 - 20
11800
67
XM_025151532.1
XP_025007300.1
21 - 22
12317
67
XM_025151533.1
XP_025007301.1
19 - 20
12167
67
XM_025151534.1
XP_025007302.1
18 - 19
2669
67
XM_025151535.1
XP_025007303.1
13 - 14
10871
67
XM_025151536.1
XP_025007304.1
10 - 11
2530
67
XM_025151537.1
XP_025007305.1
10 - 11
1667
67
XM_025151538.1
XP_025007306.1
11 - 12
4826
67
XM_025151539.1
XP_025007307.1
9 - 10
1246
67
XM_025151540.1
XP_025007308.1
10 - 11
3252
67
XM_025151541.1
XP_025007309.1
10 - 11
3233
67
XM_025151543.1
XP_025007311.1
8 - 9
910
67
XM_025151544.1
XP_025007312.1
20 - 21
12266
67
XM_025151545.1
XP_025007313.1
7 - 8
709
67
GenBank mRNA
CR391233.1
-
2 - 3
204
67
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Recommended Assay
Gallus gallus
Probe spans exons
Panther Classification:
Molecular Function -
non-motor actin binding protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0