Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAGLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416634.5 | XP_416634.1 | 1 - 2 | 201 | 70 | |
GenBank mRNA | BX929918.2 | - | 1 - 2 | 376 | 70 | |
CR407457.1 | - | 1 - 2 | 164 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |