Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAGLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015297685.2 | XP_015153171.1 | 2 - 3 | 336 | 93 | |
XM_416634.5 | XP_416634.1 | 2 - 3 | 381 | 93 | ||
GenBank mRNA | BX929918.2 | - | 2 - 3 | 556 | 93 | |
BX933937.1 | - | 1 - 2 | 322 | 93 | ||
CR407457.1 | - | 2 - 3 | 344 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |