Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM208A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015293389.2 | XP_015148875.1 | 7 - 8 | 1899 | 96 | |
XM_015293390.2 | XP_015148876.1 | 7 - 8 | 1898 | 96 | ||
XM_015293391.2 | XP_015148877.1 | 7 - 8 | 1899 | 96 | ||
XM_015293392.2 | XP_015148878.1 | 7 - 8 | 1909 | 96 | ||
XM_015293393.2 | XP_015148879.1 | 7 - 8 | 1897 | 96 | ||
XM_015293394.2 | XP_015148880.1 | 7 - 8 | 1902 | 96 | ||
XM_015293395.2 | XP_015148881.1 | 7 - 8 | 2032 | 96 | ||
XM_015293396.2 | XP_015148882.1 | 7 - 8 | 1022 | 96 | ||
XM_025154719.1 | XP_025010487.1 | 7 - 8 | 794 | 96 | ||
GenBank mRNA | BX935948.2 | - | 5 - 6 | 557 | 96 | |
CR407463.1 | - | 2 - 3 | 167 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |