Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277525.1 | NP_001264454.1 | 3 - 4 | 393 | 114 | |
GenBank mRNA | BX930426.1 | - | 3 - 4 | 393 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |