Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015284385.2 | XP_015139871.1 | 4 - 5 | 2952 | 105 | |
GenBank mRNA | BX931270.1 | - | 3 - 4 | 544 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |