Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004935622.2 | XP_004935679.1 | 1 - 2 | 257 | 118 | |
GenBank mRNA | BX932296.1 | - | 1 - 2 | 302 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |