Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003643010.4 | XP_003643058.1 | 1 - 2 | 2713 | 75 | |
GenBank mRNA | BX932659.1 | - | 1 - 2 | 210 | 75 |
Gene Symbol: CDC14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004949234.3 | XP_004949291.1 | 1 - 2 | 155 | 75 | |
XM_004949235.3 | XP_004949292.1 | 1 - 2 | 155 | 75 | ||
XM_004949236.3 | XP_004949293.1 | 1 - 2 | 155 | 75 | ||
XM_004949237.3 | XP_004949294.1 | 1 - 2 | 155 | 75 | ||
XM_015280388.2 | XP_015135874.1 | 1 - 2 | 155 | 75 | ||
XM_025144747.1 | XP_025000515.1 | 1 - 2 | 155 | 75 | ||
XM_025144750.1 | XP_025000518.1 | 1 - 2 | 155 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |