Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277175.1 | NP_001264104.1 | 2 | 252 | 80 | |
XM_025155451.1 | XP_025011219.1 | 2 | 156 | 80 | ||
GenBank mRNA | BX932967.1 | - | 1 | 51 | 80 | |
BX933093.2 | - | 2 | 182 | 80 | ||
BX934630.1 | - | 2 | 217 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |