Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP27C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004942891.3 | XP_004942948.1 | 3 - 4 | 110 | ||
XM_025152584.1 | XP_025008352.1 | 2 - 3 | 110 | |||
GenBank mRNA | BX933060.1 | - | 3 - 4 | 833 | 110 | |
CR387850.1 | - | 2 - 3 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |