Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277784.1 | NP_001264713.1 | 1 - 2 | 214 | 97 | |
XM_025149634.1 | XP_025005402.1 | 2 - 3 | 1397 | 97 | ||
GenBank mRNA | BU258980.1 | - | 1 - 2 | 222 | 97 | |
BX929847.1 | - | 1 - 2 | 222 | 97 | ||
BX934937.1 | - | 1 - 2 | 162 | 97 | ||
CR353827.1 | - | 1 - 2 | 136 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |