Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277492.1 | NP_001264421.1 | 5 - 6 | 1198 | 108 | |
GenBank mRNA | BX935008.1 | - | 5 - 6 | 1192 | 108 | |
KU180444.1 | - | 3 - 4 | 1069 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |