Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPA33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416656.6 | XP_416656.1 | 5 - 6 | 780 | 84 | |
GenBank mRNA | BX934867.1 | - | 5 - 6 | 705 | 84 | |
BX936257.1 | - | 5 - 6 | 705 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |