Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419990.3 | XP_419990.2 | 2 - 3 | 512 | 109 | |
GenBank mRNA | CR352371.1 | - | 2 - 3 | 174 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |