Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277630.1 | NP_001264559.1 | 7 - 8 | 814 | 108 | |
NM_001277631.1 | NP_001264560.1 | 6 - 7 | 694 | 108 | ||
XM_015283717.2 | XP_015139203.1 | 7 - 8 | 893 | 108 | ||
XM_025148693.1 | XP_025004461.1 | 7 - 8 | 799 | 108 | ||
GenBank mRNA | AJ444864.1 | - | 4 - 5 | 501 | 108 | |
BU224670.1 | - | 3 - 4 | 319 | 108 | ||
BU338692.1 | - | 2 - 3 | 172 | 108 | ||
BX929977.2 | - | 6 - 7 | 678 | 108 | ||
BX934152.2 | - | 7 - 8 | 815 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |