Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025148782.1 | XP_025004550.1 | 3 - 4 | 360 | 101 | |
GenBank mRNA | BX930452.2 | - | 3 - 4 | 335 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |