Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RILPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025155656.1 | XP_025011424.1 | 3 - 4 | 631 | 99 | |
GenBank mRNA | BX932368.2 | - | 3 - 4 | 607 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |