Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199656.1 | NP_001186585.1 | 1 - 2 | 241 | 141 | |
GenBank mRNA | BX933277.2 | - | 1 - 2 | 224 | 141 | |
CR353343.1 | - | 1 - 2 | 244 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |