Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277486.1 | NP_001264415.1 | 2 - 3 | 160 | 135 | |
GenBank mRNA | BX933901.2 | - | 2 - 3 | 229 | 135 | |
BX934934.1 | - | 2 - 3 | 170 | 135 | ||
CN220727.1 | - | 2 - 3 | 160 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |