Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277486.1 | NP_001264415.1 | 4 - 5 | 314 | 97 | |
GenBank mRNA | BX933901.2 | - | 4 - 5 | 383 | 97 | |
BX934934.1 | - | 4 - 5 | 324 | 97 | ||
CN220727.1 | - | 4 - 5 | 314 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |