Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIVA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277535.1 | NP_001264464.1 | 2 - 3 | 372 | 78 | |
GenBank mRNA | BX931304.1 | - | 2 - 3 | 354 | 78 | |
BX934499.2 | - | 2 - 3 | 372 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |