Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277841.1 | NP_001264770.1 | 3 - 4 | 215 | 121 | |
XM_015293127.2 | XP_015148613.1 | 3 - 4 | 387 | 121 | ||
GenBank mRNA | BX934554.2 | - | 3 - 4 | 215 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |