Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277841.1 | NP_001264770.1 | 6 - 7 | 631 | 70 | |
XM_015293127.2 | XP_015148613.1 | 6 - 7 | 803 | 70 | ||
GenBank mRNA | AJ396688.1 | - | 4 - 5 | 443 | 70 | |
BX934554.2 | - | 6 - 7 | 631 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |