Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199584.1 | NP_001186513.1 | 6 - 7 | 1163 | 74 | |
XM_015286372.2 | XP_015141858.1 | 7 - 8 | 1347 | 74 | ||
XM_015286373.2 | XP_015141859.1 | 10 - 11 | 3153 | 74 | ||
XM_025150668.1 | XP_025006436.1 | 7 - 8 | 1352 | 74 | ||
XM_025150669.1 | XP_025006437.1 | 10 - 11 | 3153 | 74 | ||
GenBank mRNA | BX934411.1 | - | 3 - 4 | 370 | 74 | |
BX936092.2 | - | 2 - 3 | 210 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |