Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTX3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015289955.2 | XP_015145441.1 | 4 - 5 | 2169 | 136 | |
XM_422114.6 | XP_422114.4 | 4 - 5 | 2149 | 136 | ||
GenBank mRNA | BX950444.1 | - | 2 - 3 | 780 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |