Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277762.1 | NP_001264691.1 | 2 - 3 | 340 | 118 | |
GenBank mRNA | BU297448.1 | - | 2 - 3 | 340 | 118 | |
BX950616.1 | - | 2 - 3 | 340 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |