Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNF4beta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CR352592.1 | - | 4 - 5 | 515 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |